Methyl-3-amino-4-methylbenzoate ≥98.0%

Supplier: TCI America

A1432-25G
TCA1432-025GEA 37.92 CAD
TCA1432-025G
Methyl-3-amino-4-methylbenzoate ≥98.0%
Methyl 3-amino-4-methylbenzoate

CAS Number: 18595-18-1
MDL Number: MFCD00025206
Molecular Formula: C9H11NO2
Molecular Weight: 165.19
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 116


Other Product Names / Synonyms: 3-Amino-4-methylbenzoic Acid Methyl Ester, Methyl 3-Amino-p-toluate, 3-Amino-p-toluic Acid Methyl Ester

Formula: C₉H₁₁NO₂
MW: 165.19
Melting Pt: 116 °C
MDL Number: MFCD00025206
CAS Number: 18595-18-1
https://ca.vwr.com/stibo/web/std.lang.all/94/25/28929425.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR