4-Amino-2,3,5,6-tetrafluoropyridine ≥97.0%

Supplier: TCI America

Synonyms: 2,3,5,6-Tetrafluoropyridin-4-amine

A1546-1G
TCA1546-001GEA 67.32 CAD
TCA1546-001G
4-Amino-2,3,5,6-tetrafluoropyridine ≥97.0%
4-Amino-2,3,5,6-tetrafluoropyridine

CAS Number: 1682-20-8
MDL Number: MFCD00010855
Molecular Formula: C5H2F4N2
Molecular Weight: 166.08
Purity/Analysis Method: >97.0% (GC)
Form: Crystal
Melting point (°C): 86

Formula: C₅H₂F₄N₂
MW: 166.08
Melting Pt: 86 °C
MDL Number: MFCD00010855
CAS Number: 1682-20-8
https://ca.vwr.com/stibo/web/std.lang.all/96/42/28929642.jpg

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR