2-Acetyl-3-methylpyrazine ≥98.0%

Supplier: TCI America

A1978-1G
TCA1978-1GEA 31.6 CAD
TCA1978-1G
2-Acetyl-3-methylpyrazine ≥98.0%
2-Acetyl-3-methylpyrazine

CAS Number: 23787-80-6
MDL Number: MFCD00014612
Molecular Formula: C7H8N2O
Molecular Weight: 136.15
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 69
Flash Point (°C): 80
Specific Gravity (20/20): 1.12

Formula: C₇H₈N₂O
MW: 136.15 g/mol
Boiling Pt: 69 °C
Density: 1.12 (20 °C)
Flash Pt: 80 °C
MDL Number: MFCD00014612
CAS Number: 23787-80-6
https://ca.vwr.com/stibo/web/std.lang.all/01/09/28930109.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR