3-Amino-2-nitropyridine ≥98.0%
Supplier: TCI America
Synonyms:
2-Nitropyridin-3-amine
, 2-Nitropyridin-3-ylamine
, 2-Nitro-3-pyridylamine
A2149-1G
TCA2149-1GEA
91.5
CAD
TCA2149-1G
3-Amino-2-nitropyridine ≥98.0%
3-Amino-2-nitropyridine
CAS Number: 13269-19-7
MDL Number: MFCD00044102
Molecular Formula: C5H5N3O2
Molecular Weight: 139.11
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 201
Formula:
C₅H₅N₃O₂ MW: 139.11 g/mol Melting Pt: 201 °C |
MDL Number:
MFCD00044102 CAS Number: 13269-19-7 |
![]() |
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...