Bis(4-bromophenyl)ether ≥97.0%

Supplier: TCI America

Synonyms: 4,4'-Dibromodiphenyl Ether4 4'-OXYBIS(BROMOBENZENE)

B0834-25G
TCB0834-25GEA 146.94 CAD
TCB0834-25G
Bis(4-bromophenyl)ether ≥97.0%
Bis(4-bromophenyl)ether

CAS Number: 2050-47-7
MDL Number: MFCD00000095
Molecular Formula: C12H8Br2O
Molecular Weight: 328.00
Purity/Analysis Method: >97.0% (GC)
Form: Crystal
Boiling point (°C): 205
Melting point (°C): 60


Product Categories: Materials Chemistry, Diphenyl Ethers (for High-Performance Polymer Research)
Other Product Names / Synonyms: 4,4'-Dibromodiphenyl Ether

Formula: C₁₂H₈Br₂O
MW: 328.00 g/mol
Boiling Pt: 205 °C
Melting Pt: 60 °C
MDL Number: MFCD00000095
CAS Number: 2050-47-7
https://ca.vwr.com/stibo/web/std.lang.all/27/41/28932741.jpg

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR