N-Benzylformamide ≥98.0% (by GC, total nitrogen)
Supplier: TCI America
Synonyms:
N-Formylbenzylamine
B2184-25G
B2184-5G
TCB2184-25GEA
195.92
CAD
TCB2184-25G
TCB2184-5G
N-Benzylformamide ≥98.0% (by GC, total nitrogen)
N-Benzylformamide
CAS Number: 6343-54-0
MDL Number: MFCD00003281
Molecular Formula: C8H9NO
Molecular Weight: 135.17
Purity/Analysis Method: >98.0% (GC,N)
Form: Crystal
Melting point (°C): 62
Other Product Names / Synonyms: N-Formylbenzylamine
Formula:
C₈H₉NO Melting Pt: 62 °C |
MDL Number:
MFCD00003281 CAS Number: 6343-54-0 |
![]() |
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...