1-Benzyl-1H-indazol-3-ol ≥98.0%
Supplier: TCI America
Synonyms:
1-Benzyl-3-hydroxy-1H-indazole
, 1-Benzyl-2,3-dihydroindazol-3-one
B2411-5G
B2411-25G
TCB2411-5GEA
39.5
CAD
TCB2411-5G
TCB2411-25G
1-Benzyl-1H-indazol-3-ol ≥98.0%
1-Benzyl-1H-indazol-3-ol
CAS Number: 2215-63-6
MDL Number: MFCD01631173
Molecular Formula: C14H12N2O
Molecular Weight: 224.26
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 167
Other Product Names / Synonyms: 1-Benzyl-2,3-dihydroindazol-3-one, 1-Benzyl-3-hydroxy-1H-indazole
Formula:
C₁₄H₁₂N₂O Melting Pt: 167 °C |
MDL Number:
MFCD01631173 CAS Number: 2215-63-6 |
![]() |
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...