2-Bromo-5-nitro-3-picoline ≥98.0%

Supplier: TCI America

Synonyms: 2-Bromo-3-methyl-5-nitropyridine

B2798-1G B2798-5G
TCB2798-1GEA 75 CAD
TCB2798-1G TCB2798-5G
2-Bromo-5-nitro-3-picoline ≥98.0%
2-Bromo-5-nitro-3-picoline

CAS Number: 23132-21-0
Molecular Formula: C6H5BrN2O2
Molecular Weight: 217.02
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 58


Other Product Names / Synonyms: 2-Bromo-5-nitro-3-picoline

Formula: C₆H₅BrN₂O₂
MW: 217.02 g/mol
Melting Pt: 58 °C
MDL Number: MFCD03095065
CAS Number: 23132-21-0
https://ca.vwr.com/stibo/web/std.lang.all/43/02/28934302.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR