3-(Benzyloxy)-4-methoxybenzaldehyde ≥97.0%

Supplier: TCI America

Synonyms: 3-Benzyloxy-4-methoxybenzaldehyde

B2885-25G
TCB2885-25GEA 254.38 CAD
TCB2885-25G
3-(Benzyloxy)-4-methoxybenzaldehyde ≥97.0%
3-(Benzyloxy)-4-methoxybenzaldehyde

CAS Number: 6346-05-0
MDL Number: MFCD00003386
Molecular Formula: C15H14O3
Molecular Weight: 242.27
Purity/Analysis Method: >97.0% (GC)
Form: Crystal
Boiling point (°C): 120
Melting point (°C): 63

Formula: C₁₅H₁₄O₃
MW: 242.27 g/mol
Boiling Pt: 120 °C
Melting Pt: 63 °C
MDL Number: MFCD00003386
CAS Number: 6346-05-0
https://ca.vwr.com/stibo/web/std.lang.all/43/67/28934367.jpg

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR