4-Bromo-2,5-difluoroanisole ≥98.0%

Supplier: TCI America

B3097-5G
TCB3097-5GEA 82.5 CAD
TCB3097-5G
4-Bromo-2,5-difluoroanisole ≥98.0%
4-Bromo-2,5-difluoroanisole

CAS Number: 202865-60-9
MDL Number: MFCD00143258
Molecular Formula: C7H5BrF2O
Molecular Weight: 223.02
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 63


Product Categories: Synthetic Organic Chemistry, Fluorinated Building Blocks
Other Product Names / Synonyms: 1-Bromo-2,5-difluoro-4-methoxybenzene

Formula: C₇H₅BrF₂O
MW: 223.02 g/mol
Melting Pt: 63 °C
MDL Number: MFCD00143258
CAS Number: 202865-60-9
https://ca.vwr.com/stibo/web/std.lang.all/45/37/28934537.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR