Chlorazol black E
Supplier: TCI America
Synonyms:
Direct Black 38
C0533-25G
TCC0533-025GEA
91.64
CAD
TCC0533-025G
Chlorazol black E
Chlorazol black E
CAS Number: 1937-37-7
MDL Number: MFCD00066363
Molecular Formula: C34H27N9O7S2
Molecular Weight: 781.73
Form: Crystal
Color: Black
Other Product Names / Synonyms: Direct Black 38
Formula:
C₃₄H₂₇N₉O₇S₂ MW: 781.73 g/mol |
MDL Number:
MFCD00066363 CAS Number: 1937-37-7 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...