4-Chlorophenoxyacetyl chloride ≥98.0% (by titrimetric analysis)

Supplier: TCI America

C0832-100G C0832-25G C0832-500G
TCC0832-100GEA 360.24 CAD
TCC0832-500G TCC0832-25G TCC0832-100G
4-Chlorophenoxyacetyl chloride ≥98.0% (by titrimetric analysis)
4-Chlorophenoxyacetyl chloride

CAS Number: 4122-68-3
MDL Number: MFCD00000727
Molecular Formula: C8H6Cl2O2
Molecular Weight: 205.03
Purity/Analysis Method: >98.0% (T)
Form: Clear Liquid
Boiling point (°C): 165
Flash Point (°C): 113
Freezing point (°C): 18
Specific Gravity (20/20): 1.37

Formula: C₈H₆Cl₂O₂
Boiling Pt: 165 °C
Density: 1.37 (20 °C)
Flash Pt: 113 °C
MDL Number: MFCD00000727
CAS Number: 4122-68-3
https://ca.vwr.com/stibo/web/std.lang.all/79/23/28937923.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR