1-Chloroisoquinoline ≥98.0%

Supplier: TCI America

C2097-25G C2097-5G
TCC2097-5GEA 151.68 CAD
TCC2097-5G TCC2097-25G
1-Chloroisoquinoline ≥98.0%
1-Chloroisoquinoline

CAS Number: 19493-44-8
MDL Number: MFCD00024134
Molecular Formula: C9H6ClN
Molecular Weight: 163.60
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Boiling point (°C): 108
Melting point (°C): 36
Flash Point (°C): 110
Storage Temperature: 0-10°C

Formula: C₉H₆ClN
MW: 163.61 g/mol
Boiling Pt: 108 °C
Melting Pt: 36 °C
Flash Pt: 110 °C
MDL Number: MFCD00024134
CAS Number: 19493-44-8
https://ca.vwr.com/stibo/web/std.lang.all/91/52/28939152.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR