N,N'-Diphenylformamidine ≥98.0% (by GC, titration analysis)
Supplier: TCI America
D1221-25G
D1221-250G
TCD1221-025GEA
71.1
CAD
TCD1221-025G
TCD1221-250G
N,N'-Diphenylformamidine ≥98.0% (by GC, titration analysis)
N,N'-Diphenylformamidine
CAS Number: 622-15-1
MDL Number: MFCD00003026
Molecular Formula: C13H12N2
Molecular Weight: 196.25
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Melting point (°C): 138
Formula:
C₁₃H₁₂N₂ MW: 196.25 g/mol Melting Pt: 138 °C |
MDL Number:
MFCD00003026 CAS Number: 622-15-1 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...