Evans blue
Supplier: TCI America
E0197-25G
TCE0197-025GEA
126.29
CAD
TCE0197-025G
Evans blue
Evans blue
CAS Number: 314-13-6
MDL Number: MFCD00004021
Molecular Formula: C34H28N6O14S4
Molecular Weight: 960.79
Form: Crystal
Color: Deep Yellow Red
Formula:
C₃₄H₂₈N₆O₁₄S₄ |
MDL Number:
MFCD00004021 CAS Number: 314-13-6 |
![]() |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...