D-(-)-α-(4-Hydroxyphenyl)glycine ≥99.0% (by HPLC, titration analysis)

Supplier: TCI America

Synonyms: H-D-Phg(4-OH)-OH(-)-4-Hydroxy-D-phenylglycine(R)-(-)-2-(4-Hydroxyphenyl)glycineD-(-)-p-HydroxyphenylglycineD(-)-4-Hydroxyphenylglycine

H0758-25G H0758-100G H0758-500G
TCH0758-100GEA 121.66 CAD
TCH0758-025G TCH0758-500G TCH0758-100G
D-(-)-α-(4-Hydroxyphenyl)glycine ≥99.0% (by HPLC, titration analysis)
D(-)-α-(4-Hydroxyphenyl)glycine

CAS Number: 22818-40-2
MDL Number: MFCD00004262
Molecular Formula: C8H9NO3
Molecular Weight: 167.16
Purity/Analysis Method: >99.0% (HPLC,T)
Form: Crystal
Melting point (°C): 240
Specific rotation [a]20/D: -158 deg (C=1, 1mol/L HCl)


Product Categories: Synthetic Organic Chemistry, Chiral Building Blocks, Bioscience, non-Proteinogenic Amino Acids
Other Product Names / Synonyms: (R)-(-)-2-Amino-2-(4-hydroxyphenyl)acetic Acid, D-(-)-2-(4-Hydroxyphenyl)glycine, H-D-Phg(4-OH)-OH

https://ca.vwr.com/stibo/web/std.lang.all/07/92/28970792.jpg

Order Now

Specification Test Results

Purity/Analysis Method 99.0% (HPLC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR