5-Methylisophthalic acid ≥98.0% (by GC, titration analysis)
Supplier: TCI America
M1903-5G
M1903-25G
TCM1903-25GEA
208.56
CAD
TCM1903-25G
TCM1903-5G
5-Methylisophthalic acid ≥98.0% (by GC, titration analysis)
5-Methylisophthalic acid
CAS Number: 499-49-0
MDL Number: MFCD00013986
Molecular Formula: C9H8O4
Molecular Weight: 180.16
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Melting point (°C): 298
Formula:
C₉H₈O₄ Melting Pt: 298 °C |
MDL Number:
MFCD00013986 CAS Number: 499-49-0 |
![]() |
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...