Phenylpyruvic acid ≥93.0% (by GC, titration analysis)
Supplier: TCI America
Synonyms:
alpha-Keto-DL-phenylalanine
P0154-5G
P0154-25G
TCP0154-005GEA
123.4
CAD
TCP0154-005G
TCP0154-025G
Phenylpyruvic acid ≥93.0% (by GC, titration analysis)
Phenylpyruvic acid
CAS Number: 156-06-9
MDL Number: MFCD00002589
Molecular Formula: C9H8O3
Molecular Weight: 164.16
Purity/Analysis Method: >93.0% (GC,T)
Form: Crystal
Storage Temperature: <0°C
Other Product Names / Synonyms: alpha-Keto-DL-phenylalanine
Formula:
C₉H₈O₃ MW: 164.16 g/mol |
MDL Number:
MFCD00002589 CAS Number: 156-06-9 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...