N,N-Dimethyl-4-(2-pyridylazo)aniline ≥98.0% (by HPLC, titration analysis)

Supplier: TCI America

P0617-1G
TCP0617-001GEA 263.86 CAD
TCP0617-001G
N,N-Dimethyl-4-(2-pyridylazo)aniline ≥98.0% (by HPLC, titration analysis)
N,N-Dimethyl-4-(2-pyridylazo)aniline

CAS Number: 13103-75-8
MDL Number: MFCD00010435
Molecular Formula: C13H14N4
Molecular Weight: 226.28
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal
Melting point (°C): 110


Other Product Names / Synonyms: 2-(4-Dimethylaminophenylazo)pyridine, 4-(2-Pyridylazo)-N,N-dimethylaniline

Formula: C₁₃H₁₄N₄
Melting Pt: 110 °C
MDL Number: MFCD00010435
CAS Number: 13103-75-8
https://ca.vwr.com/stibo/web/std.lang.all/13/45/28991345.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR