2-Carboxyphenyl salicylate ≥98.0% (by HPLC, titration analysis)
Supplier: TCI America
Synonyms:
Salicylsalicylic acid
, O-Salicyloylsalicylic Acid
, Salicylic Acid 2-Carboxyphenyl Ester
, 2-Salicyloyloxybenzoic Acid
, Salsalate
CAS Number: 552-94-3
MDL Number: MFCD00020252
Molecular Formula: C14H10O5
Molecular Weight: 258.23
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal
Color: White
Melting point (°C): 137
Other Product Names / Synonyms: 2-Salicyloyloxybenzoic Acid, O-Salicyloylsalicylic Acid, Salicylic Acid 2-Carboxyphenyl Ester, Salsalate
Formula:
C₁₄H₁₀O₅ MW: 258.23 g/mol Melting Pt: 137 °C |
MDL Number:
MFCD00020252 CAS Number: 552-94-3 |
![]() |
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...