Tetrabutylammonium hydroxide 10% in methanol
Supplier: TCI America
T0717-25ML
T0717-500ML
T0717-100ML
TCT0717-100MLEA
79
CAD
TCT0717-500ML
TCT0717-100ML
TCT0717-25ML
Tetrabutylammonium hydroxide 10% in methanol
Tetrabutylammonium hydroxide
CAS Number: 2052-49-5
MDL Number: MFCD00009425
Molecular Formula: C16H37NO
Molecular Weight: 259.48
Form: Clear Liquid
Color: Colorless
Flash Point (°C): 11
Specific Gravity (20/20): 0.82
Formula:
C₁₆H₃₇NO MW: 259.48 g/mol Density: 0.82 (20 °C) Flash Pt: 11 °C |
MDL Number:
MFCD00009425 CAS Number: 2052-49-5 UN: 3263 ADR: 8,II |
![]() |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...