2,3,5,6-Tetramethylpyrazine ≥98.0% (by GC, titration analysis)

Supplier: TCI America

T0972-25G
TCT0972-25GEA 53.31 CAD
TCT0972-25G
2,3,5,6-Tetramethylpyrazine ≥98.0% (by GC, titration analysis)
2,3,5,6-Tetramethylpyrazine

CAS Number: 1124-11-4
MDL Number: MFCD00006146
Molecular Formula: C8H12N2
Molecular Weight: 136.20
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Color: White
Boiling point (°C): 190
Melting point (°C): 86
Flash Point (°C): 100

Formula: C₈H₁₂N₂
MW: 136.2 g/mol
Boiling Pt: 190 °C
Melting Pt: 86 °C
Flash Pt: 100 °C
MDL Number: MFCD00006146
CAS Number: 1124-11-4
https://ca.vwr.com/stibo/web/std.lang.all/50/57/28995057.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR