Tetrakis(dimethylsiloxy)silane ≥96.0%

Supplier: TCI America

T2102-5G
TCT2102-5GEA 48.98 CAD
TCT2102-5G
Tetrakis(dimethylsiloxy)silane ≥96.0%
Tetrakis(dimethylsiloxy)silane

CAS Number: 17082-47-2
MDL Number: MFCD00053848
Molecular Formula: C8H28O4Si5
Molecular Weight: 328.73
Purity/Analysis Method: >96.0% (GC)
Form: Clear Liquid
Color: Colorless
Boiling point (°C): 190
Melting point (°C): -60
Flash Point (°C): 67
Specific Gravity (20/20): 0.89


Other Product Names / Synonyms: Tetrakis(dimethylsilyl) Orthosilicate

Formula: C₈H₂₈O₄Si₅
Boiling Pt: 190 °C
Melting Pt: –60 °C
Density: 0.89 (20 °C)
Flash Pt: 67 °C
MDL Number: MFCD00053848
CAS Number: 17082-47-2
https://ca.vwr.com/stibo/web/std.lang.all/59/98/28995998.jpg

Order Now

Specification Test Results

Purity/Analysis Method 96.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR