1H-Tridecafluorohexane ≥97.0%

Supplier: TCI America

Synonyms: 1H-Perfluorohexane

T2496-5G
TCT2496-5GEA 197.5 CAD
TCT2496-5G
1H-Tridecafluorohexane ≥97.0%
1H-Tridecafluorohexane

CAS Number: 355-37-3
MDL Number: MFCD00142632
Molecular Formula: C6HF13
Molecular Weight: 320.05
Purity/Analysis Method: >97.0% (GC)
Form: Clear Liquid
Color: Colorless
Boiling point (°C): 70
Specific Gravity (20/20): 1.69


Product Categories: Synthetic Organic Chemistry, Fluorous Compounds
Other Product Names / Synonyms: 1H-Perfluorohexane

Formula: C₆HF₁₃
MW: 320.05 g/mol
Boiling Pt: 70 °C
Density: 1.69 (20 °C)
MDL Number: MFCD00142632
CAS Number: 355-37-3
https://ca.vwr.com/stibo/web/std.lang.all/62/26/28996226.jpg

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR