4-(Trimethylsilyl)diphenylacetylene ≥97.0%
Supplier: TCI America
Synonyms:
Trimethyl[4-(phenylethynyl)phenyl]silane
T2596-5G
TCT2596-5GEA
251.22
CAD
TCT2596-5G
4-(Trimethylsilyl)diphenylacetylene ≥97.0%
4-(Trimethylsilyl)diphenylacetylene
CAS Number: 136459-72-8
MDL Number: MFCD09953470
Molecular Formula: C17H18Si
Molecular Weight: 250.42
Purity/Analysis Method: >97.0% (GC)
Form: Crystal
Color: White
Melting point (°C): 39
Storage Temperature: 0-10°C
Other Product Names / Synonyms: Trimethyl[4-(phenylethynyl)phenyl]silane
Formula:
C₁₇H₁₈Si Melting Pt: 39 °C |
MDL Number:
MFCD09953470 CAS Number: 136459-72-8 |
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...