Poly(styrene) 2,5% (w/w) dispersion in water, latex microsphere 6.0 micron
Supplier: Thermo Scientific Chemicals
42716-AA
42716-AA
CAAA42716-AAEA
883.62
CAD
CAAA42716-AA
AA42716-AA
Poly(styrene) 2,5% (w/w) dispersion in water, latex microsphere 6.0 micron
Poly(styrene)
Formula:
(C₈H₈)n MW: 125,000-250,000 Storage Temperature: Refrigerator |
MDL Number:
MFCD00084450 CAS Number: 9003-53-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...