Poly(styrene) 2,5% (w/w) dispersion in water, latex microsphere 75.0 micron
Supplier: Thermo Scientific Chemicals
42750-AA
42750-AA
CAAA42750-AAEA
878.76
CAD
CAAA42750-AA
AA42750-AA
Poly(styrene) 2,5% (w/w) dispersion in water, latex microsphere 75.0 micron
Poly(styrene)
Formula:
(C₈H₈)n MW: 125,000-250,000 Storage Temperature: Freezer |
MDL Number:
MFCD00084450 CAS Number: 9003-53-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...