1,1'-Ferrocenedicarboxaldehyde ≥97%

44292-02 44292-04 44292-04 44292-02
CAAAA44292-04EA 1374.24 CAD
AAA44292-02 AAA44292-04 CAAAA44292-04 CAAAA44292-02
1,1'-Ferrocenedicarboxaldehyde ≥97%
1,1'-Ferrocenedicarboxaldehyde

Powder

Formula: C₁₂H₁₀FeO₂
MW: 242.06 g/mol
Melting Pt: 183…184 °C
Storage Temperature: Ambient
MDL Number: MFCD02093629
CAS Number: 1271-48-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR