Fullerene buckytube/nanotube single-walled >60%

Supplier: Thermo Scientific Chemicals
Warning

44501-02 44501-77 44501-02 44501-77
CAAAA44501-77EA 459.12 CAD
CAAAA44501-77 CAAAA44501-02 AAA44501-77 AAA44501-02
Fullerene buckytube/nanotube single-walled >60%
Fullerene buckytube/nanotube

Powder

Storage Temperature: Ambient
MDL Number: MFCD00282904
CAS Number: 308068-56-6
UN: 1325
ADR: 4.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR