Polyphenylmethylsiloxane, M.W. 2500-2700

Supplier: Thermo Scientific Chemicals

44569-18 44569-A1 44569-A1 44569-30 44569-18 44569-30
CAAAA44569-30EA 260.08 CAD
AAA44569-18 CAAAA44569-30 CAAAA44569-A1 AAA44569-A1 CAAAA44569-18 AAA44569-30
Polyphenylmethylsiloxane, M.W. 2500-2700
Polyphenylmethylsiloxane

Liquid

Formula: (C₆H₅(CH₃)SiO₂)₂
Flash Pt: 300 °C (572 °F)
Storage Temperature: Ambient
MDL Number: MFCD00146731
CAS Number: 9005-12-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR