2-Fluoroisonicotinic acid 98%
Supplier: Thermo Scientific Chemicals
Synonyms:
2-Fluoropyridine-4-carboxylic acid
44755-03
CAAAA44755-01EA
224.11
CAD
CAAAA44755-01
CAAAA44755-03
2-Fluoroisonicotinic acid 98%
2-Fluoroisonicotinic acid
Powder
Purity: 98%
CAS No.: 402-65-3
Molecular Formula: C6H4FNO2
Formula Weight: 141.10
Physical Form: Powder
Melting Point: 195-497°
MDL No.: MFCD02181194
Formula:
C₆H₄FNO₂ MW: 141.10 g/mol Melting Pt: 195…497 °C Storage Temperature: Ambient |
MDL Number:
MFCD02181194 CAS Number: 402-65-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...