Nickel A-5000, sponge nonpromoted
Supplier: VWR
|
Danger
|
44867-36
44867-22
44867-36
44867-22
CAAAA44867-22EA
264.21
CAD
AAA44867-36
CAAAA44867-22
CAAAA44867-36
AAA44867-22
Nickel A-5000, sponge nonpromoted
Nickel
Slurry in water
Applications: Hydrogenation of nitro groups to amines, hydrogenation of carbonyl groups, hydrogenation of olefins to alkanes, reductive alkylation (stereoselective)
Formula:
Ni MW: 58.69 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00011137 CAS Number: 7440-02-0 UN: 1378 ADR: 4.2,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...