Cobalt A-8B46, sponge promoted with Nickel and Chromium

Danger

44871-36 44871-36 44871-22 44871-22
CAAAA44871-22EA 255.03 CAD
CAAAA44871-22 AAA44871-22 AAA44871-36 CAAAA44871-36
Cobalt A-8B46, sponge promoted with Nickel and Chromium
Cobalt

Slurry in water


Sponge Cobalt, A-8B46, promoted with Nickel and Chromium, is used in synthetic chemistry for hydrogenation reactions.

Formula: Co
MW: 58.93 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00239945
CAS Number: 7440-48-4
UN: 1378
ADR: 4.2,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR