TBE (10X), TRIS + borate + EDTA, Thermo Fisher Scientific

Supplier: Thermo Fisher Scientific

J62449-K3 J62449-K7 J62449-K2 J62449-K2 J62449-K3 J62449-K7
CAAAJ62449-K3EA 102.75 CAD
CAAAJ62449-K3 AAJ62449-K7 AAJ62449-K2 CAAAJ62449-K2 AAJ62449-K3 CAAAJ62449-K7
TBE (10X), TRIS + borate + EDTA, Thermo Fisher Scientific
Buffers Electrophoresis Buffers Running Buffers
Physical Form: Liquid
Appearance: Colorless
MDL No.: MFCD00236358
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR