IEF Anode buffer (50X), Thermo Fisher Scientific

Supplier: Thermo Fisher Scientific

J63303-K2 J63303-K2 J63303-AP J63303-AP
CAAAJ63303-K2EA 85.59 CAD
AAJ63303-K2 CAAAJ63303-K2 CAAAJ63303-AP AAJ63303-AP
IEF Anode buffer (50X), Thermo Fisher Scientific
Buffers Electrophoresis Buffers Running Buffers
For isoelectric focusing (IEF)

Storage Temperature: 2°C to 8°C
Physical Form: Liquid
Appearance: Colorless
MDL No.: MFCD03846400
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR