TRIS-Tricine-SDS running buffer (10X), anode buffer, pH 8.9, Thermo Fisher Scientific

Supplier: Thermo Fisher Scientific
J63375-K2 J63375-AP J63375-AP J63375-K2
CAAAJ63375-K2EA 82.28 CAD
CAAAJ63375-K2 CAAAJ63375-AP AAJ63375-AP AAJ63375-K2
TRIS-Tricine-SDS running buffer (10X), anode buffer, pH 8.9, Thermo Fisher Scientific
Buffers Electrophoresis Buffers Running Buffers
Liquid

Physical Form: Liquid
Applications: For TRIS-Tricine gel electrophoresis
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR