TAE (10X), TRIS + acetate + EDTA, Thermo Fisher Scientific

Supplier: Thermo Fisher Scientific
J63677-K3 J63677-K3 J63677-K2 J63677-K2 J63677-K7 J63677-K7
CAAAJ63677-K3EA 102.75 CAD
AAJ63677-K3 CAAAJ63677-K3 CAAAJ63677-K2 AAJ63677-K2 AAJ63677-K7 CAAAJ63677-K7
TAE (10X), TRIS + acetate + EDTA, Thermo Fisher Scientific
Buffers Electrophoresis Buffers Running Buffers
Physical Form: Liquid
MDL No.: MFCD00236357
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR