  • Supplier
  • Refine by Suppliers
    Sort by:
  • Search Within Results
Torque Meters
https://ca.vwr.com/stibo/smallweb/std.lang.all/99/28/18989928.jpg
Portable torque meters are used for applying torque to bolts, for torque wrench calibration, switch or contact closure measurement, and other industrial, QC, and materials testing applications. Torque meters are easy-to-use handheld units that can be used to record the torque in rotating systems in a variety of settings. These meters feature multiple units of measure as well as features such as hold, peak, min/max, fast/slow sampling, push button high/low-resolution selection, auto power off, and user-friendly computer interface.
SearchPresentationType-VERTICAL
SearchResultCount:"1"
  List View Easy View
Sort by:
Supplier:  Sper Scientific
Description:   This portable meter is used for bolt torqueing, torque wrench calibration, switch or contact closure measurement and other industrial, QC, and materia...
Call for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at 1-800-932-5000.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at 1-800-932-5000.
You must log in to order restricted items. We request that you provide the required business documentation to purchase this product for the first time.
To order chemicals, medical devices, or other restricted products please provide identification that includes your business name and shipping address via email CMD_NA@vwr.com or fax 484.881.5997 referencing your VWR account number . Acceptable forms of identification are:
  • issued document with your organization's Federal Tax ID Number
  • Government issued document with your organization's Resale Tax ID Number
  • Any other Government ID that includes the business name and address
VWR will not lift restrictions for residential shipping addresses.
???store.product.restricted???
This product has been blocked by your organization. Please contact your purchasing department for more information.
The original product is no longer available. The replacement shown is available.
This product is currently unavailable but limited stock may be available in our extended warehouse network. Please call 1-800-932-5000 and a VWR Customer Service Representative will help you.