  • Supplier
  • Refine by Suppliers
    Sort by:
  • Search Within Results
Radio Frequency Meters
https://ca.vwr.com/stibo/smallweb/std.lang.all/97/02/18989702.jpg
Radio frequency is the range of electromagnetic frequencies above the audio range and below infrared light, and these radio frequency meters are useful for evaluating the frequencies in this range emitted by welding equipment, microwave oven leakage, and other environmental safety concerns. These meters feature maximum and minimum hold, audible alarms, and a zero adjustment option to eliminate background EMF. Displays also indicate overload and low battery.
SearchPresentationType-VERTICAL
SearchResultCount:"1"
  List View Grid View Easy View (new)
Sort by:
Catalog Number: (97049-648)
Supplier:  Sper Scientific
Description:   Evaluate the RF emissions of welding equipment, microwave oven leakage, and other environmental safety concerns.
Call for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at 1-800-932-5000.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at 1-800-932-5000.
You must log in to order restricted items. We request that you provide the required business documentation to purchase this product for the first time.
To order chemicals, medical devices, or other restricted products please provide identification that includes your business name and shipping address via email CMD_NA@vwr.com or fax 484.881.5997 referencing your VWR account number . Acceptable forms of identification are:
  • issued document with your organization's Federal Tax ID Number
  • Government issued document with your organization's Resale Tax ID Number
  • Any other Government ID that includes the business name and address
VWR will not lift restrictions for residential shipping addresses.
???store.product.restricted???
This product has been blocked by your organization. Please contact your purchasing department for more information.
The original product is no longer available. The replacement shown is available.
This product is currently unavailable but limited stock may be available in our extended warehouse network. Please call 1-800-932-5000 and a VWR Customer Service Representative will help you.
1 - 1  of 1