Nɑ-(tert-Butoxycarbonyl)-D-asparagine

Supplier: BACHEM AMERICAS INC

Synonyms: Boc-D-Asn-OHNɑ-Boc-D-asparagine

4002271.0005
A-1220.0005BAEA 198.84 CAD
A-1220.0005BA
Nɑ-(tert-Butoxycarbonyl)-D-asparagine
Nalpha-(tert-Butoxycarbonyl)-D-asparagine

Sequence: Boc-D-Asn-OH


For isoasparagine derivatives please see the subfamily "Isoasparagine" filed under "Aspartic Acid".

Formula: C₉H₁₆N₂O₅
Storage Temperature: Ambient
MDL Number: MFCD00065558
CAS Number: 75647-01-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR