Boc-Ser(tBu)-OH
Supplier: Bachem Americas
Certificates
About this item
Synonyms: Boc-Ser(tBu)-OH,N-Boc-O-tert-butyl-L-serine
Formula:C₁₂H₂₃NO₅
MW:261.31 g/mol
MDL Number:MFCD00079666
CAS Number:13734-38-8
:Length:
:Width:
:Height:
:Ø
:Thickness:
:Boiling Pt:
:Boiling Pt:
:Melting Pt:
:Flash Pt:
:Flash Pt:
:M.W. 261.31 g/mol
Storage Temperature:Freezer
Customers Who Bought This Also Bought