1-Benzyl-N-carbobenzoxy-D-glutamate

Supplier: BACHEM AMERICAS INC

Synonyms: Z-D-Glu-OBzl1-Benzyl N-Cbz-D-glutamateN-Cbz-D-glutamic Acid 1-Benzyl EsterN-Carbobenzoxy-D-glutamic Acid 1-Benzyl EsterN-CBZ-D-glutamic acid alpha-benzyl ester

4007080.0005
C-3120.0005BAEA 434.04 CAD
C-3120.0005BA
1-Benzyl-N-carbobenzoxy-D-glutamate
1-Benzyl-N-carbobenzoxy-D-glutamate

Sequence: Z-D-Glu-OBzl


The derivatives of Glu-NH₂ are assembled in the "Isoglutamine" subfamily.

Formula: C₂₀H₂₁NO₆
Storage Temperature: Ambient
MDL Number: MFCD00069647
CAS Number: 65706-99-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR