N(α)-Acetyl-D-tryptophan

Supplier: BACHEM AMERICAS INC

Synonyms: Ac-D-Trp-OH

4027317.0005
F-1090.0005BAEA 186.03 CAD
F-1090.0005BA
N(α)-Acetyl-D-tryptophan
N(α)-Acetyl-D-tryptophan

Sequence: Ac-D-Trp-OH

Formula: C₁₃H₁₄N₂O₃
Storage Temperature: Ambient
MDL Number: MFCD00065021
CAS Number: 2280-01-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR