3-(2-Naphthyl)-D-alanine
Catalog #: F-1860.0001BA
Supplier: Bachem Americas
Price
$493.64
Log in for availability
Specifications
- Size:1 g
- Size:1 g
- Cat. no.:F-1860.0001BA
Specifications
Specification Test Results
- 3-(2-Naphthyl)-D-alanine:
Specification Test Results
About this item
Synonyms: (R)-alpha-Amino-2-naphthalenepropionic Acid,H-D-2-Nal-OH
:Length:
:Width:
:Height:
:Ø
:Thickness:
:Boiling Pt:
:Boiling Pt:
:Melting Pt:
:Flash Pt:
:Flash Pt:
:M.W. 215.25 g/mol
Formula:C₁₃H₁₃NO₂
MW:215.25 g/mol
Storage Temperature:Ambient
MDL Number:MFCD00038546
CAS Number:76985-09-6
Customers Who Bought This Also Bought