Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?
  • Criteria
  • Supplier
  • Refine by Suppliers
    Sort by:

  • Sale Items
You Searched For:

Pocket Glass Thermometers

8  results were found
SearchPresentationType-VERTICAL
 
 
SearchResultCount:"8"
  List View Easy View
Sort by:
 
 
 
 

Supplier:  VWR International
Description:   Pocket glass thermometers feature permanently fused markings.
Supplier:  VWR International
Description:   Featuring a closed metal case to protect from breakage, these total immersion thermometers are a compact 160mm (6.3”), making them ideal for field stu...
Supplier:  VWR International
Description:   Glass pocket thermometers are conveniently sized and accurate to within one degree.
Supplier:  Bel-Art Products
Description:   EnviroKleen™ certified thermometers feature green, non-toxic, biodegradable, Enviro-Safe® liquid against lead-free, white back glass
Product and/or packaging is safer to human and environmental health
Supplier:  Bel-Art Products
Description:   Featuring a case to protect from breakage, these total immersion thermometers are a compact 160mm (6.3”), making them ideal for field studies, waste w...
Supplier:  Bel-Art Products
Description:   Equipped with a case to protect from breakage, these total immersion thermometers are ideal for field studies, waste water treatment and any “on-the-g...
Product and/or packaging is safer to human and environmental health
Catalog Number: (470018-442)

Supplier:  Kemtec
Description:   Microscale Thermometers, Range: 0–100°C
Catalog Number: (470104-652)

Supplier:  Kemtec
Description:   Microchemistry Thermometers
Call for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at 1-800-932-5000.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at 1-800-932-5000.
You must log in to order restricted items. We request that you provide the required business documentation to purchase this product for the first time.

To order chemicals, medical devices, or other restricted products please provide identification that includes your business name and shipping address via email CMD_NA@vwr.com or fax 484.881.5997 referencing your VWR account number . Acceptable forms of identification are:

  • issued document with your organization's Federal Tax ID Number
  • Government issued document with your organization's Resale Tax ID Number
  • Any other Government ID that includes the business name and address


VWR will not lift restrictions for residential shipping addresses.

-Additional Documentation May be needed to purchase this item. A VWR representative will contact you if needed.
This product has been blocked by your organization. Please contact your purchasing department for more information.
The original product is no longer available. The replacement shown is available.
This product is currently unavailable but limited stock may be available in our extended warehouse network. Please call 1-800-932-5000 and a VWR Customer Service Representative will help you.
1 - 8  of 8