Would you like to log in again?
CFM/CMM Thermo–Anemometer with Built–in InfraRed Thermometer, Extech
Supplier: Extech
Built–in IR Thermometer measures non–contact surface temperature. Model HD300 is a InfraRed Thermometer measures remote surface temperatures to 932°F (500°C) with 30:1 distance to spot ratio and Laser pointer. It features a simultaneous display of Air Flow in CFM/CMM or Air Velocity plus Ambient Temperature. It also features easy to set Area dimensions (cm2) that are stored in the meter’s internal memory for the next power on.
Certifications: CE marked.
Ordering information: 3 year warranty. Includes Windows® compatible software with cable, vane sensor with 3.9ft (120cm) cable, hard carrying case, and 9V battery.
Log in to see your contract pricing and availability.
  Remember me on this device
ORDER
Expand All / Collapse All
Description Supplier No. VWR Catalog Number Unit Price Quantity
CFM/CMM Thermo–Anemometer with built–in InfaRed Thermometer with certificate of calibration traceable to NIST HD300-NIST 10171-852 Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
SPECIFICATIONS
Temperature Accuracy 2ºC (±4ºF)
Temperature range 14 to 140ºF, –10 to 60ºC
Air velocity range 80 to 5900 ft/min, 0.4 to 30.00 m/s, 1.4 to 108.0 km/h, 0.9 to 67.0 MPH, 0.8 to 58.0 knots
Air velocity accuracy ±3%
Air flow range 0 to 999900
IR Temperature Range –58 to 932ºF, –50 to 500ºC
IR Temperature Accuracy ±2%
Enter Details
Select the article for which you are looking for Certificate
Indicate the Lot Number. This field is mandatory.
Enter Certificate Details
Select Product:
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty