Anti-PCSK6 Rabbit Polyclonal Antibody

Supplier: Enzo Life Sciences
210-517-R100
89145-844EA 566.22 CAD
89145-844
Anti-PCSK6 Rabbit Polyclonal Antibody
Antibodies
Host: Rabbit

  • For Immunoprecipitation, Western Blot

Recognizes human PACE4.

Type: Primary
Antigen: PCSK6
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR