Phalloidin, amino-, tosylate ≥90% (by HPLC)
Supplier: Enzo Life Sciences
|
Danger
|
350-266-M001
89149-370EA
1806.15
CAD
89149-370
Phalloidin, amino-, tosylate ≥90% (by HPLC)
Phalloidin, amino-, tosylate
Actin ligand.
Biological activity like phalloidin, but contains a functional amino group for coupling reactions.
Soluble in DMSO, dimethyl formamide, methanol or water.
Formula:
C₃₅H₄₉N₉O₁₀S·C₇H₇O₃S Storage Temperature: Freezer |
UN:
3462 ADR: 6.1,I |
Specification Test Results
Purity | ≥90% (HPLC) |
Source | Semisynthetic from phalloidin |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...