(+)-Taxifolin ≥90% (by HPLC)
Supplier: Enzo Life Sciences
385-018-M050
385-018-M010
89150-520EA
297.1
CAD
89150-520
89150-518
(+)-Taxifolin ≥90% (by HPLC)
(+)-Taxifolin
Anti-inflammatory and chemopreventive
- Formulation: Light yellow to brown powder.
Antioxidant flavonoid. Anti-inflammatory compound. Chemopreventive agent. Decreases hepatic lipid synthesis.
Formula:
C₁₅H₁₂O₇ |
MDL Number:
MFCD00006845 CAS Number: 480-18-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...