Aminopterin

Supplier: Enzo Life Sciences

440-041-M050
89151-336EA 157.37 CAD
89151-336
Aminopterin
Aminopterin

Folic acid antagonist. Potent dihydrofolate reductase inhibitor.

Formula: C₁₉H₂₀N₈O₅
MW: 440.42 g/mol
MDL Number: MFCD00075621
CAS Number: 54-62-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR